Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02581573_m1
          See other ODAM GE Assays ›
          Gene Symbol:
          Odam
          Gene Name:
          odontogenic, ameloblast asssociated
          Gene Aliases:
          2310011G06Rik, APIN
          Chromosome Location:
          Chr.5: 87885025 - 87894174 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02581573_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Odam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027128.2 NP_081404.2 8 - 9 659 122
          GenBank mRNA AK009298.1 - 5 - 6 363 122
          BC132394.1 - 5 - 6 303 40130671 122
          BC132396.1 - 5 - 6 302 40130673 122
          EF113909.1 - 8 - 9 659 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000800.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Odam

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          biomineral tissue development
          odontogenesis of dentin-containing tooth
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502685 AK009298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502685 AK009298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963758 EF113909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963758 NM_027128.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557635 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67451 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170654 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502685 AK009298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502685 AK009298.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963758 EF113909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963758 NM_027128.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23870106 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05645826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170654 - -
          Whole Mouse Genome G4122A A_51_P502685 AK009298.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502685 AK009298.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04538439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04641865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04545646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04641870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23870123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04545646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04547393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04641863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04547400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04547391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04538440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04403203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04538443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23870152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10522882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522881 - -
          Affymetrix HT MG-430 PM Array Plate 1430186_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10522875 - -
          Mouse Genome 430 2.0 Array 1430186_at - -
          Mouse Genome 430 2.0 Array 1430186_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69592_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69592_at - -
          Affymetrix Mouse Clariom S Array PSR0500007029.mm.1 TC0500000800.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007029.mm.1 NM_027128.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007029.mm.1 NM_027128.2 Refseq
          Mouse Expression Set 430 1430186_PM_at - -
          Mouse Expression Set 430 1430186_at - -
          Affymetrix HT Mouse Genome 430B Array 1430186_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430186_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659342 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline