Search
Search

Clec4b2| Gene Symbol: Clec4b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004159.2 | NP_001004159.1 | 3 - 4 | 244 | 101 | |
| GenBank mRNA | AK156711.1 | - | 3 - 4 | 376 | 101 | |
| AY365133.1 | - | 3 - 4 | 267 | 101 | ||
Set Membership: |
|