| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700025775.mm.1 |
TC0700002884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025775.mm.1 |
NR_073038.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025774.mm.1 |
TC0700002884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025774.mm.1 |
NR_073038.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025771.mm.1 |
TC0700002884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025771.mm.1 |
NR_073038.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025772.mm.1 |
TC0700002884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025772.mm.1 |
NR_073038.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700025773.mm.1 |
TC0700002884.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700025773.mm.1 |
NR_073038.1 |
Refseq |