Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033263.5 | NP_001028435.1 | 5 - 6 | 2249 | 59 | |
NM_001301014.1 | NP_001287943.1 | 5 - 6 | 2249 | 59 | ||
GenBank mRNA | AK147446.1 | - | 5 - 6 | 2250 | 59 | |
AK220325.1 | - | 5 - 6 | 1124 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |