| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900016878.mm.1 |
NM_001033325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016878.mm.1 |
NM_001033325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016878.mm.1 |
TC0900002034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016880.mm.1 |
NM_001033325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016880.mm.1 |
NM_001033325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016880.mm.1 |
TC0900002034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016882.mm.1 |
NM_001033325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016882.mm.1 |
NM_001033325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016882.mm.1 |
TC0900002034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016881.mm.1 |
NM_001033325.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016881.mm.1 |
NM_001033325.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016881.mm.1 |
TC0900002034.mm.1 |
MTA TC |