| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2422650 |
AK017143.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011647 |
BC050254.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011647 |
AK139013.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2011647 |
BC026499.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1959560 |
AK139013.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
281683 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
790857 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
441803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692393 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729741 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
195282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
816882 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
AK014301.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
AK136286.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
BC057012.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
BC057013.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
BC078417.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
BC132116.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
BC132118.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
K02583.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
NM_007776.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438865 |
NM_007777.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3778 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
110669 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
97279 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
690560 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
40669 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
18351 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798102 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
890603 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921853 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
966777 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
210446 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836761 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10265 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
180113 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
170440 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1959560 |
AK139013.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2422650 |
AK017143.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011647 |
BC050254.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011647 |
AK139013.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2011647 |
BC026499.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05504423 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05504406 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05504379 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05504354 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P217492 |
AK017143.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05504406 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536294 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536398 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04496135 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536291 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536391 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04380182 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536397 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536182 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536212 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04714306 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536415 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536210 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536295 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536178 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04714308 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536219 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04566072 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536384 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04350252 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536395 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04380178 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536389 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536393 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536216 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536289 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536400 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536284 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04496137 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536373 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536401 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536223 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536380 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536217 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536176 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536185 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04956687 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536409 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536288 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536405 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536221 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536174 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536407 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04355270 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536209 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536403 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536377 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536290 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04909715 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536215 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536220 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04711368 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536387 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536214 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536222 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23536286 |
- |
- |