Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033421.3 | NP_001028593.1 | 1 - 2 | 95 | 71 | |
GenBank mRNA | AK136651.1 | - | 1 - 2 | 96 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |