Search
Search

Grxcr2| Gene Symbol: Grxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033426.2 | NP_001028598.1 | 1 - 2 | 385 | 125 | |
| GenBank mRNA | AK158418.1 | - | 1 - 2 | 386 | 125 | |
| AK158432.1 | - | 270 | 125 | |||
| BC147102.1 | - | 1 - 2 | 380 | 9056155 | 125 | |
| BC147103.1 | - | 1 - 2 | 380 | 9056135 | 125 | |
Set Membership: |
|