Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601151_g1
          See other D930028M14RIK GE Assays ›
          Gene Symbol:
          D930028M14Rik
          Gene Name:
          RIKEN cDNA D930028M14 gene
          Gene Aliases:
          EG434147
          Chromosome Location:
          Chr.7: 25152457 - 25156630 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601151_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D930028M14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045847.1 2 - 3 463 67
          NR_045848.1 2 - 3 361 67
          GenBank mRNA AK142847.1 - 2 - 3 464 67
          BE456889.1 - 2 - 3 361 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000406.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P744437 AK142847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P744437 AK142847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P744437 AK142847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P744437 AK142847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P744437 AK142847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P744437 AK142847.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P06024353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06076831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06255644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06593012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06344448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06344454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06400219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06344430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06344419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05932983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25008658 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1457086_PM_at - -
          Mouse Genome 430 2.0 Array 1457086_PM_at - -
          Mouse Genome 430 2.0 Array 1457086_at - -
          Affymetrix Mouse Clariom S Array PSR0700003079.mm.1 TC0700000406.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003079.mm.1 NR_045848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003079.mm.1 NR_045848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003080.mm.1 TC0700000406.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003080.mm.1 NR_045847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003080.mm.1 NR_045847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003080.mm.1 NR_045848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003080.mm.1 NR_045848.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003078.mm.1 TC0700000406.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003078.mm.1 NR_045847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003078.mm.1 NR_045847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003081.mm.1 TC0700000406.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003081.mm.1 NR_045847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003081.mm.1 NR_045847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003081.mm.1 NR_045848.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003081.mm.1 NR_045848.1 Refseq
          Mouse Expression Set 430 1457086_PM_at - -
          Mouse Expression Set 430 1457086_at - -
          Murine Genome U74v2 Set 169817_at - -
          Murine Genome U74v2 Set 137358_at - -
          Murine Genome U74v2 Set 137358_PM_at - -
          Murine Genome U74v2 Set 169817_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457086_at - -
          Affymetrix HT Mouse Genome 430B Array 1457086_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0