Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601400_m1
          See other DEFB36 GE Assays ›
          Gene Symbol:
          Defb36
          Gene Name:
          defensin beta 36
          Gene Aliases:
          BD-36, Defb123, mBD-36
          Chromosome Location:
          Chr.2: 152604327 - 152612729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601400_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037247.4 NP_001032324.1 1 - 2 222 92
          GenBank mRNA AF533211.1 - 1 - 2 82 92
          AK005872.1 - 1 - 2 177 92
          AY591385.1 - 1 - 2 86 92
          DQ012042.1 - 1 - 2 222 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002291.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Mus musculus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          biological_process
          defense response to bacterium
          innate immune response
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 NM_001037247.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AF533211.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AK005872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AY591385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 DQ012042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 NM_001037247.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AF533211.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AK005872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 AY591385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58990 DQ012042.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064506 NM_001037247.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064506 AK005872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064506 AY591385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064506 DQ012042.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701707 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50837 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154100 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 NM_001037247.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AF533211.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AK005872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AY591385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 DQ012042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 NM_001037247.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AF533211.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AK005872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 AY591385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58990 DQ012042.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064506 NM_001037247.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064506 AK005872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064506 AY591385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064506 DQ012042.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750143 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00528774 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154100 - -
          Whole Mouse Genome G4122A A_52_P58990 NM_001037247.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AF533211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AK005872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AY591385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 DQ012042.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 NM_001037247.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AF533211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AK005872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 AY591385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58990 DQ012042.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113870 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01493688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01200957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01977659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01848126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02034073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02057235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02034071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01977651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01977661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477137 - -
          Affymetrix Mouse Gene 1.1 ST Array 10477134 - -
          Affymetrix Mouse Clariom S Array PSR0200017916.mm.1 NM_001037247.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017916.mm.1 NM_001037247.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017916.mm.1 TC0200002291.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017914.mm.1 NM_001037247.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017914.mm.1 NM_001037247.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017914.mm.1 TC0200002291.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017915.mm.1 NM_001037247.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017915.mm.1 NM_001037247.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017915.mm.1 TC0200002291.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017913.mm.1 NM_001037247.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017913.mm.1 NM_001037247.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017913.mm.1 TC0200002291.mm.1 MTA TC
          Murine Genome U74v2 Set 170425_at - -
          Murine Genome U74v2 Set 170425_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702382 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2926781 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702382 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2926781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline