| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2140418 |
BY721060.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2140418 |
DQ012044.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2140418 |
NM_001037751.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2140418 |
AK136677.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128809 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
662871 |
NM_019398.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
764631 |
AK015642.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274262 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402808 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250654 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2140418 |
BY721060.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2140418 |
DQ012044.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2140418 |
NM_001037751.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2140418 |
AK136677.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08136285 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08136285 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658635 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658593 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658580 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658532 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658620 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658565 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658560 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658518 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658582 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658554 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658572 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658515 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658598 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658513 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658567 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658615 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658562 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658606 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658600 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658537 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658547 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658621 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658534 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658549 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658539 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658508 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658563 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658542 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658579 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658570 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658568 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658630 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29658611 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11812645 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658515 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658594 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658572 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658524 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658590 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658591 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11586574 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658584 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658578 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658570 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658589 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658523 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11285751 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658556 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658560 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658569 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658518 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11289640 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658577 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658520 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11295587 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11368065 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658557 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11499520 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658565 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658516 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658615 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11295586 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658617 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11289634 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658579 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11180751 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658609 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658598 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658568 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658607 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658582 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658602 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658513 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658575 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658585 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658586 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658562 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658567 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11368072 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658603 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658583 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29658593 |
- |
- |