Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601703_g1
          See other GNG5 GE Assays ›
          Gene Symbol:
          Gng5
          Gene Name:
          guanine nucleotide binding protein (G protein), gamma 5
          Gene Aliases:
          G(y)5
          Chromosome Location:
          Chr.3: 146499836 - 146505543 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601703_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gng5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010318.2 NP_034448.2 1 - 2 181 95
          GenBank mRNA AK146065.1 - 1 - 2 104 95
          AK204635.1 - 1 - 2 26 95
          BC002316.1 - 1 - 2 132 3593324 95
          BC010725.1 - 1 - 2 127 95
          BC087872.1 - 1 - 2 103 6582824 95
          BY132164.1 - 1 - 2 181 95
          U38498.1 - 1 - 2 56 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gng5

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of secondary heart field cardioblast proliferation
          positive regulation of neural precursor cell proliferation
          PDZ domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22614973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22614975 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75038 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107738 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988259 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237166 BC010725.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237166 BC087872.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237166 NM_010318.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237166 AK146065.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237166 BC002316.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22614988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22615014 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107738 - -
          Whole Mouse Genome G4122A A_52_P640337 BC010725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678261 BC010725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678261 BC087872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678261 U38498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678261 NM_010318.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678261 AK146065.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237166 BC010725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237166 BC087872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237166 NM_010318.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237166 AK146065.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237166 BC002316.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124765 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125406 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131809 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03089392 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03089381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22614973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22614975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22614984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22615066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03089388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22614999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22615006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10496823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434674 - -
          Affymetrix HT MG-430 PM Array Plate 1423985_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10496822 - -
          Affymetrix Mouse Gene 1.1 ST Array 10434672 - -
          Mouse Genome 430 2.0 Array 1423985_at - -
          Mouse Genome 430 2.0 Array 1436946_s_at - -
          Mouse Genome 430 2.0 Array 1436946_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423985_PM_at - -
          Mouse Genome 430 2.0 Array 1425012_PM_at - -
          Mouse Genome 430 2.0 Array 1425012_at - -
          Affymetrix Mouse Clariom S Array PSR0300012100.mm.1 NM_010318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012100.mm.1 NM_010318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012100.mm.1 TC0300001493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012102.mm.1 NM_010318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012102.mm.1 NM_010318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012102.mm.1 TC0300001493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012097.mm.1 NM_010318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012097.mm.1 NM_010318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012097.mm.1 TC0300001493.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300012104.mm.1 NM_010318.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012104.mm.1 NM_010318.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300012104.mm.1 TC0300001493.mm.1 MTA TC
          Mouse Expression Set 430 1423985_at - -
          Mouse Expression Set 430 1423985_PM_at - -
          Mouse Expression Set 430 1436946_PM_s_at - -
          Mouse Expression Set 430 1425012_PM_at - -
          Mouse Expression Set 430 1425012_at - -
          Mouse Expression Set 430 1436946_s_at - -
          Murine Genome U74v2 Set 94042_PM_f_at - -
          Murine Genome U74v2 Set 94042_f_at - -
          Mouse Genome 430A 2.0 Array 1436946_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1423985_at - -
          Mouse Genome 430A 2.0 Array 1436946_s_at - -
          Mouse Genome 430A 2.0 Array 1423985_PM_at - -
          Mouse Genome 430A 2.0 Array 1425012_at - -
          Mouse Genome 430A 2.0 Array 1425012_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893304 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610403 - -
          Illumina mouse-6 v1.0 expression beadchip 50112 - -
          Illumina mouse-6 v1.0 expression beadchip 610403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163159 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253827 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740257 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740257 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163159 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253827 - -
          mouse-6 v1.1 (Illumina) GI_84579905-A - -
          mouse-6 v1.1 (Illumina) scl014707.2_63-S - -
          mouse-6 v1.1 (Illumina) scl22478.3_284-S - -
          Illumina Sentrix mouse-6.v1 bead array 610403 - -
          Illumina Sentrix mouse-6.v1 bead array 50112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014707.2_63-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22478.3_284-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610403 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163159 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163159 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253827 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_84579905-A - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline