Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601831_g1
          See other RPS26 GE Assays ›
          Gene Symbol:
          Rps26
          Gene Name:
          ribosomal protein S26
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 128624529 - 128626506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601831_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013765.2 NP_038793.2 3 - 4 346 83
          GenBank mRNA AK008301.1 - 3 - 4 342 83
          AK010689.2 - 3 - 4 341 83
          AK012722.1 - 3 - 4 344 83
          AK018702.1 - 3 - 4 342 83
          AK152132.1 - 3 - 4 342 83
          AV585706.1 - 2 - 3 300 83
          BC036987.1 - 2 - 3 332 4987659 83
          BC081452.1 - 2 - 3 319 6411109 83
          BC100456.1 - 3 - 4 333 6541446 83
          BU530648.1 - 3 - 4 347 83
          U67770.1 - 2 - 3 323 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps26

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of RNA splicing
          mRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27542346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27542347 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2184246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302087 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2184246 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302675 NM_001178001.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302675 BC096641.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302675 BU530648.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302675 U67770.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302675 BC100456.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27542348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121113 - -
          Whole Mouse Genome G4122A A_51_P302675 NM_001178001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302675 BC096641.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302675 BU530648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302675 U67770.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302675 BC100456.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120557 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106676 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08640155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08640165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08640164 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08640157 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08640167 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27542346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27542347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08640152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08477593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08367138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08405907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08350470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08429147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08367136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08443389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08751817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08247058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08210551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08754159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27542315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08477591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08657666 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373500 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373501 - -
          Affymetrix HT MG-430 PM Array Plate 1415876_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581558 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549375 - -
          Affymetrix Mouse Gene 1.1 ST Array 10373498 - -
          Mouse Genome 430 2.0 Array 1415876_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415876_a_at - -
          Affymetrix Mouse Clariom S Array PSR1000023753.mm.1 NM_013765.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023753.mm.1 NM_013765.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023753.mm.1 TC1000003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023754.mm.1 NM_013765.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023754.mm.1 NM_013765.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023754.mm.1 TC1000003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023752.mm.1 NM_013765.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023752.mm.1 NM_013765.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023752.mm.1 TC1000003155.mm.1 MTA TC
          Mouse Expression Set 430 1415876_PM_a_at - -
          Mouse Expression Set 430 1415876_a_at - -
          Murine Genome U74v2 Set 98565_PM_at - -
          Murine Genome U74v2 Set 98565_at - -
          Murine Genome U74v2 Set 98564_PM_f_at - -
          Murine Genome U74v2 Set 98564_f_at - -
          Murine Genome U74v2 Set 137217_f_at - -
          Murine Genome U74v2 Set 137217_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1415876_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1415876_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903111 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 510139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510139 - -
          mouse-6 v1.1 (Illumina) scl027370.3_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 510139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl027370.3_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2794676 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2794676 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746234 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl027370.3_85-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline