Search
Search

Hnrnpa3| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053263.1 | NP_444493.1 | 4 - 5 | 545 | 144 | |
| NM_146130.3 | NP_666242.2 | 4 - 5 | 611 | 144 | ||
| NM_198090.2 | NP_932758.1 | 4 - 5 | 611 | 144 | ||
| GenBank mRNA | AB201711.1 | - | 4 - 5 | 591 | 144 | |
| AF463524.1 | - | 4 - 5 | 523 | 144 | ||
| AK133860.1 | - | 4 - 5 | 610 | 144 | ||
| AK169663.1 | - | 4 - 5 | 593 | 144 | ||
| BC023828.1 | - | 4 - 5 | 564 | 5343619 | 144 | |
| BC023908.1 | - | 4 - 5 | 580 | 5322033 | 144 | |
| BC024454.1 | - | 1 - 2 | 128 | 144 | ||
| BC038364.1 | - | 4 - 5 | 593 | 5362605 | 144 | |
| BC057655.1 | - | 4 - 5 | 524 | 144 | ||
| BC062198.1 | - | 4 - 5 | 602 | 2655879 | 144 | |
| BC064824.1 | - | 4 - 5 | 576 | 6477672 | 144 | |
| BC085474.1 | - | 3 - 4 | 486 | 30538435 | 144 | |
| BC092083.1 | - | 1 - 2 | 206 | 144 | ||
| BC116250.1 | - | 4 - 5 | 514 | 40093600 | 144 | |
| BC116251.1 | - | 4 - 5 | 514 | 40093602 | 144 | |
| BC157919.1 | - | 4 - 5 | 514 | 9007347 | 144 | |
| BC158038.1 | - | 4 - 5 | 514 | 9007341 | 144 | |
| CO045618.1 | - | 3 - 4 | 395 | 144 | ||
Set Membership: |
|