Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02604120_g1
          See other ZFP93 GE Assays ›
          Gene Symbol:
          Zfp93
          Gene Name:
          zinc finger protein 93
          Gene Aliases:
          Znf235
          Chromosome Location:
          Chr.7: 24270418 - 24277794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02604120_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009567.4 NP_033593.1 4 2319 134
          GenBank mRNA AK028905.1 - 4 2157 134
          AK040325.1 - 4 2250 134
          AK132229.1 - 4 2320 134
          AK146095.1 - 4 2310 134
          AK165630.1 - 4 2274 134
          BC003776.1 - 2042 3496816 134
          BC061487.1 - 3 2100 5349153 134
          U46186.1 - 4 2311 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp93

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424561 U46186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424561 BC003776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424561 AK146095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424561 AK040325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424561 AK132229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972560 AK132229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972560 NM_009567.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049138 U46186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049138 BC003776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049138 AK146095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049138 AK040325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049138 AK132229.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136634 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972560 AK132229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972560 NM_009567.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424561 U46186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424561 BC003776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424561 AK146095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424561 AK040325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424561 AK132229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049138 U46186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049138 BC003776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049138 AK146095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049138 AK040325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049138 AK132229.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424558 U46186.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424558 BC003776.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424558 BC061487.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424558 AK146095.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424558 AK040325.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25004042 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129562 - -
          Whole Mouse Genome G4122A A_51_P424558 U46186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424558 BC003776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424558 BC061487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424558 AK146095.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424558 AK040325.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130248 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06378512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200832 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06378513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25004046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06466857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06137453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25004051 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550867 - -
          Affymetrix HT MG-430 PM Array Plate 1418955_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550865 - -
          Mouse Genome 430 2.0 Array 1441876_x_at - -
          Mouse Genome 430 2.0 Array 1418955_at - -
          Mouse Genome 430 2.0 Array 1418955_PM_at - -
          Mouse Genome 430 2.0 Array 1441876_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22755_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22755_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002732.mm.1 TC0700000376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002732.mm.1 NM_009567.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002732.mm.1 NM_009567.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002724.mm.1 TC0700000376.mm.1 MTA TC
          Mouse Expression Set 430 1418955_PM_at - -
          Mouse Expression Set 430 1418955_at - -
          Mouse Expression Set 430 1441876_PM_x_at - -
          Mouse Expression Set 430 1441876_x_at - -
          Murine Genome U74v2 Set 92990_PM_at - -
          Murine Genome U74v2 Set 92990_at - -
          Mouse Genome 430A 2.0 Array 1418955_at - -
          Mouse Genome 430A 2.0 Array 1418955_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418955_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418955_at - -
          Affymetrix HT Mouse Genome 430B Array 1441876_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441876_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 850616 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5220068 - -
          MouseWG-6 v2 Expression BeadChip 6660528 - -
          Illumina mouse-6 v1.0 expression beadchip 1980546 - -
          Illumina mouse-6 v1.0 expression beadchip 5220068 - -
          Illumina mouse-6 v1.0 expression beadchip 6660528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005220068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232697 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2498609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2487453 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2487453 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232697 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2498609 - -
          mouse-6 v1.1 (Illumina) scl022755.4_47-S - -
          mouse-6 v1.1 (Illumina) scl0022755.1_133-S - -
          mouse-6 v1.1 (Illumina) scl022755.4_143-S - -
          Illumina Sentrix mouse-6.v1 bead array 5220068 - -
          Illumina Sentrix mouse-6.v1 bead array 1980546 - -
          Illumina Sentrix mouse-6.v1 bead array 6660528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022755.4_47-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022755.1_133-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022755.4_143-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5220068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2487453 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232697 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2498609 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2487453 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2498609 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232697 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline