Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02605425_s1
          See other LCE3F GE Assays ›
          Gene Symbol:
          Lce3f
          Gene Name:
          late cornified envelope 3F
          Gene Aliases:
          2310002A05Rik
          Chromosome Location:
          Chr.3: 92992226 - 92993426 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02605425_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lce3f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001018079.1 NP_001018089.1 2 468 140
          GenBank mRNA AK009081.1 - 1 320 140
          BC060281.1 - 2 468 30288382 140

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lce3f

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152447 BC060281.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152447 NM_001018079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152447 AK009081.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812401 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154914 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152447 BC060281.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152447 NM_001018079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152447 AK009081.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22334060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154914 - -
          Whole Mouse Genome G4122A A_52_P40244 BC060281.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P40244 NM_001018079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P40244 AK009081.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154914 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02602914 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02602918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02602915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02495050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02602923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02602929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22334129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02643614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02318946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02698568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02564473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02337567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02337573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02337575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02337566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02698569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03166756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02602926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22334099 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493909 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493908 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493903 - -
          Affymetrix Mouse Gene 1.1 ST Array 10493907 - -
          Mouse Genome 430 2.0 Array 1456248_PM_at - -
          Mouse Genome 430 2.0 Array 1456248_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69520_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69520_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006443.mm.1 NM_001018079.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006443.mm.1 NM_001018079.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006443.mm.1 TC0300000838.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006444.mm.1 NM_001018079.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006444.mm.1 NM_001018079.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006444.mm.1 TC0300000838.mm.1 MTA TC
          Mouse Expression Set 430 1456248_PM_at - -
          Mouse Expression Set 430 1456248_at - -
          Murine Genome U74v2 Set 165956_PM_f_at - -
          Murine Genome U74v2 Set 165956_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2610594 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2610594 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0