Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02605693_g1
          See other ZFP114 GE Assays ›
          Gene Symbol:
          Zfp114
          Gene Name:
          zinc finger protein 114
          Gene Aliases:
          Gm1957, Zfp110
          Chromosome Location:
          Chr.7: 24175021 - 24183170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02605693_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029933.2 NP_001025104.2 4 1918 125
          GenBank mRNA AK139475.1 - 3 2837 125
          BC096771.1 - 2 2144 125
          BC138311.1 - 2 2033 8861331 125
          BC138312.1 - 2 2033 8861332 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp114

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056055 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106434 BC138312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106434 BC096771.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106434 BC138311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106434 AK139475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106434 NM_001029933.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175164 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056055 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106434 BC138312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106434 BC096771.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106434 BC138311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106434 AK139475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106434 NM_001029933.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279888 BC138312.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279888 BC096771.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279888 BC138311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279888 AK139475.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P279888 AY166793.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003761 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175164 - -
          Whole Mouse Genome G4122A A_52_P71447 BC138312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P71447 BC096771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P71447 BC138311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279888 BC138312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279888 BC096771.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279888 BC138311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279888 AK139475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P279888 AY166793.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175164 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05931923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06203167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05931922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06226851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06161700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06226849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06540196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06198088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06203166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06660401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06525960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06650523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06431277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06430469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06166975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06226850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550857 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498796 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550855 - -
          Affymetrix Mouse Gene 1.1 ST Array 10498795 - -
          Mouse Genome 430 2.0 Array 1457185_at - -
          Mouse Genome 430 2.0 Array 1457185_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002700.mm.1 TC0700000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002703.mm.1 TC0700000371.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 432880 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1580435 - -
          MouseWG-6 v2 Expression BeadChip 1230373 - -
          MouseWG-6 v2 Expression BeadChip 7000524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2454018 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446620 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219334 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2454018 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446620 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline