Search
Search

C3ar1| Gene Symbol: C3ar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009779.2 | NP_033909.1 | 2 | 3290 | 120 | |
| GenBank mRNA | AK080984.1 | - | 1 | 650 | 120 | |
| AK081010.1 | - | 1617 | 120 | |||
| U77460.1 | - | 2 | 3138 | 120 | ||
Set Membership: |
|