Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02620446_s1
          See other GPER1 GE Assays ›
          Gene Symbol:
          Gper1
          Gene Name:
          G protein-coupled estrogen receptor 1
          Gene Aliases:
          6330420K13Rik, CMKRL2, Ceprl, FEG-1, GPCR-Br, Gper, Gpr30
          Chromosome Location:
          Chr.5: 139422972 - 139427800 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02620446_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gper1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029771.3 NP_084047.2 3 2384 144
          GenBank mRNA AK018203.1 - 3 2288 144
          AK030375.1 - 3 2385 144
          CX204119.1 - 1 358 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Mouse Chemokine Panel Mouse Estrogen Panel 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gper1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          immune system process
          negative regulation of leukocyte activation
          transcription from RNA polymerase II promoter
          apoptotic process
          inflammatory response
          cell cycle
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          nervous system development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell differentiation
          apoptotic chromosome condensation
          nuclear fragmentation involved in apoptotic nuclear change
          positive regulation of cell migration
          intracellular steroid hormone receptor signaling pathway
          positive regulation of cAMP biosynthetic process
          mineralocorticoid receptor signaling pathway
          positive regulation of insulin secretion
          positive regulation of inositol trisphosphate biosynthetic process
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          steroid hormone mediated signaling pathway
          positive regulation of MAPK cascade
          innate immune response
          negative regulation of fat cell differentiation
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          positive regulation of neurogenesis
          negative regulation of DNA metabolic process
          negative regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of uterine smooth muscle contraction
          negative regulation of cell cycle arrest
          cellular response to glucose stimulus
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          cellular response to mineralocorticoid stimulus
          cellular response to estradiol stimulus
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of endothelial cell apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway
          chromatin binding
          steroid hormone receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          steroid binding
          drug binding
          mineralocorticoid receptor activity
          estrogen receptor activity
          hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 AK030375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 CX204119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 NM_029771.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 AK030375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 CX204119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 NM_029771.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110567 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 AK030375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 CX204119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 NM_029771.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 AK018203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 AK030375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 CX204119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 NM_029771.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 AK030375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 CX204119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 NM_029771.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 AK030375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 CX204119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 NM_029771.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24135531 - -
          Whole Mouse Genome G4122A A_51_P185939 AK030375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 CX204119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 NM_029771.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 AK018203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 AK030375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 CX204119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 NM_029771.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185939 AK018203.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04268252 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04268253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04268260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04268261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04268263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24135525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04802368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04802366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04271474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04268261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04955671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04748838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04112272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04748828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04591319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04112277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04112279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04112273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04112278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04436274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04271476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04748846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04802365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04268253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24135524 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526955 - -
          Affymetrix HT MG-430 PM Array Plate 1428250_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447583_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526952 - -
          Mouse Genome 430 2.0 Array 1447583_x_at - -
          Mouse Genome 430 2.0 Array 1428250_PM_at - -
          Mouse Genome 430 2.0 Array 1447583_PM_x_at - -
          Mouse Genome 430 2.0 Array 1428250_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76854_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76854_at - -
          Affymetrix Mouse Clariom S Array PSR0500015466.mm.1 TC0500001675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015466.mm.1 NM_029771.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015466.mm.1 NM_029771.3 Refseq
          Mouse Expression Set 430 1447583_x_at - -
          Mouse Expression Set 430 1447583_PM_x_at - -
          Mouse Expression Set 430 1428250_PM_at - -
          Mouse Expression Set 430 1428250_at - -
          Affymetrix HT Mouse Genome 430B Array 1447583_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1428250_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447583_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1428250_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687357 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3780014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747135 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747135 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2747135 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline