Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02620530_s1
          See other GPR84 GE Assays ›
          Gene Symbol:
          Gpr84
          Gene Name:
          G protein-coupled receptor 84
          Gene Aliases:
          EX33
          Chromosome Location:
          Chr.15: 103308233 - 103311242 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02620530_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030720.1 NP_109645.1 2 1425 139
          GenBank mRNA AF272948.1 - 2 1425 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr84

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044932 NM_030720.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044932 AF272948.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044932 BC023249.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783850 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117882 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124377 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106463 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044932 NM_030720.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044932 AF272948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2044932 BC023249.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276993 NM_030720.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P276993 AF272948.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30503397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124377 - -
          Whole Mouse Genome G4122A A_51_P276993 NM_030720.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276993 AF272948.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P141002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124377 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106463 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11880994 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12330070 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11958137 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12150583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12150582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11958138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30503406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12337454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12150582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11897993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12157825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12150583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12382125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11989322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11958136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12330070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12337453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30503396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433102 - -
          Affymetrix HT MG-430 PM Array Plate 1420591_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433101 - -
          Mouse Genome 430 2.0 Array 1420591_PM_at - -
          Mouse Genome 430 2.0 Array 1420591_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80910_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80910_at - -
          Mouse Expression Set 430 1420591_at - -
          Mouse Expression Set 430 1420591_PM_at - -
          Mouse Genome 430A 2.0 Array 1420591_at - -
          Mouse Genome 430A 2.0 Array 1420591_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420591_at - -
          Affymetrix HT Mouse Genome 430A Array 1420591_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 602240 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1212695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212695 - -
          mouse-6 v1.1 (Illumina) scl46660.3_175-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46660.3_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2991255 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1212695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991255 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212695 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46660.3_175-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline