Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02620679_s1
          See other MRGPRA3 GE Assays ›
          Gene Symbol:
          Mrgpra3
          Gene Name:
          MAS-related GPR, member A3
          Gene Aliases:
          Mrga3
          Chromosome Location:
          Chr.7: 47588950 - 47601372 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02620679_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrgpra3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153067.2 NP_694707.2 2 853 139
          GenBank mRNA AY042193.1 - 2 868 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003059.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrgpra3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          signal transduction
          G-protein coupled receptor signaling pathway
          chemosensory behavior
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417815 NM_153067.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417815 NM_153067.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87947 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417815 NM_153067.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417815 AY042193.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417815 NM_153067.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417815 NM_153067.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417815 NM_153067.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06188400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06188416 - -
          Whole Mouse Genome G4122A A_51_P417815 NM_153067.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417815 AY042193.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417815 NM_153067.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417815 AY042193.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137652 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06188416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06156602 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06156601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06346978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06108147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563724 - -
          Affymetrix HT MG-430 PM Array Plate 1426121_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563722 - -
          Mouse Genome 430 2.0 Array 1426121_PM_at - -
          Mouse Genome 430 2.0 Array 1426121_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233222_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233222_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700027969.mm.1 NM_153067.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027969.mm.1 NM_153067.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027969.mm.1 TC0700003059.mm.1 MTA TC
          Mouse Expression Set 430 1426121_at - -
          Mouse Expression Set 430 1426121_PM_at - -
          Mouse Genome 430A 2.0 Array 1426121_at - -
          Mouse Genome 430A 2.0 Array 1426121_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426121_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426121_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 786441 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 450390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226770 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226770 - -
          mouse-6 v1.1 (Illumina) scl31330.2.1_273-S - -
          Illumina Sentrix mouse-6.v1 bead array 450390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31330.2.1_273-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226770 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226770 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline