Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02620869_s1
          See other GPR21 GE Assays ›
          Gene Symbol:
          Gpr21
          Gene Name:
          G protein-coupled receptor 21
          Gene Aliases:
          C230004C13Rik
          Chromosome Location:
          Chr.2: 37516626 - 37519281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02620869_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177383.4 NP_796357.1 2 2172 160
          GenBank mRNA AK048682.1 - 2 2173 160
          AK139361.1 - 2 2175 160
          AK141999.1 - 2207 160

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr21

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          positive regulation of multicellular organism growth
          glucose homeostasis
          negative regulation of insulin receptor signaling pathway
          molecular_function
          signal transducer activity
          G-protein coupled receptor activity
          G-protein coupled amine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103147 AK139361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103147 AK141999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103147 NM_177383.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103147 AK048682.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137465 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103147 AK139361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103147 AK141999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103147 NM_177383.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103147 AK048682.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21142506 - -
          Whole Mouse Genome G4122A A_52_P420715 AY255598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P420715 BC112377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P420715 AK139361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P420715 AK141999.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P420715 NM_177383.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131486 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103147 AK139361.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103147 AK141999.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103147 NM_177383.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103147 AK048682.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01678730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01167261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01678719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01167254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01167255 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01167256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21142518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02119345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02233122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02091937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01396538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01209286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01678713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01933862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21142533 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471818 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471814 - -
          Affymetrix Mouse Clariom S Array PSR0200006101.mm.1 NM_177383.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006101.mm.1 NM_177383.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006101.mm.1 TC0200000686.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 634101 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520152 - -
          Illumina mouse-6 v1.0 expression beadchip 6520152 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520152 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667865 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667865 - -
          mouse-6 v1.1 (Illumina) scl21001.2_655-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520152 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21001.2_655-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246464 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246464 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21001.2_655-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline