Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02621016_s1
          See other GPR160 GE Assays ›
          Gene Symbol:
          Gpr160
          Gene Name:
          G protein-coupled receptor 160
          Gene Aliases:
          1700025D19Rik, GPCR150
          Chromosome Location:
          Chr.3: 30855930 - 30897194 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02621016_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr160
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134385.2 NP_001127857.1 4 2119 143
          NM_001134386.2 NP_001127858.1 3 1602 143
          NM_001286994.1 NP_001273923.1 4 2128 143
          NM_001286995.1 NP_001273924.1 3 1831 143
          NM_001286996.1 NP_001273925.1 4 1702 143
          NM_027965.2 NP_082241.1 4 1931 143
          GenBank mRNA AK154620.1 - 3 1604 143
          AK154784.1 - 4 2086 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030512 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956627 AK154620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956627 AK154784.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956627 AK006330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956627 NM_001134385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956627 NM_001134386.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433383 AK044927.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81778 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12372 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110001 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132782 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956627 AK154620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956627 AK154784.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956627 AK006330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956627 NM_001134385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956627 NM_001134386.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110001 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04862162 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04862147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132782 - -
          Whole Mouse Genome G4122A A_51_P264227 AK154620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264227 AK154784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264227 AK006330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264227 NM_001134385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P264227 NM_001134386.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22030508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03101482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02610154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02340463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02588299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02906770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02906769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03032989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03112604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02887516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03032988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22030532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03008336 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10491276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491278 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10491274 - -
          Affymetrix HT MG-430 PM Array Plate 1453072_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10491272 - -
          Mouse Genome 430 2.0 Array 1453072_at - -
          Mouse Genome 430 2.0 Array 1453072_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71862_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71862_at - -
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_001134385.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_001134386.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_001286994.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_001286995.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_001286996.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 NM_027965.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300001121.mm.1 TC0300000205.mm.1 MTA TC
          Mouse Expression Set 430 1453072_PM_at - -
          Mouse Expression Set 430 1453072_at - -
          Affymetrix HT Mouse Genome 430B Array 1453072_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453072_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884862 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621649 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621649 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline