Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02744887_s1
          See other SYNA GE Assays ›
          Gene Symbol:
          Syna
          Gene Name:
          syncytin a
          Gene Aliases:
          Gm453, Gm52, syncytin-A
          Chromosome Location:
          Chr.5: 134557254 - 134577057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02744887_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syna
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013751.2 NP_001013773.1 1 1851 73
          GenBank mRNA BC138919.1 - 1 1851 8861941 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Gene Ontology Categories:

          Function(s)

          syncytium formation by plasma membrane fusion
          labyrinthine layer development

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011814 BC138919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011814 NM_001013751.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339197 AK014261.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339197 XM_906193.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339197 XM_978146.1 refseq_mrna_mouse_XM
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011814 BC138919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011814 NM_001013751.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32983184 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534302 - -
          Affymetrix Mouse Gene 1.1 ST Array 10534301 - -
          Affymetrix Mouse Clariom S Array PSR0500030426.mm.1 TC0500003375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030426.mm.1 NM_001013751.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030426.mm.1 NM_001013751.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030425.mm.1 TC0500003375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030425.mm.1 NM_001013751.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030425.mm.1 NM_001013751.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160198 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160198 - -
          mouse-6 v1.1 (Illumina) GI_85701451-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160198 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline