| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500008276.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008276.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008276.mm.1 |
TC1500001045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500008277.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008277.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008277.mm.1 |
TC1500001045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500008275.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008275.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008275.mm.1 |
TC1500001045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500008280.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008280.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008280.mm.1 |
TC1500001045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500008278.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008278.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008278.mm.1 |
TC1500001045.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500008279.mm.1 |
NR_040351.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008279.mm.1 |
NR_040351.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500008279.mm.1 |
TC1500001045.mm.1 |
MTA TC |