| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600020200.mm.1 |
TC0600002604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600020200.mm.1 |
NM_001034902.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600020200.mm.1 |
NM_001034902.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600020201.mm.1 |
TC0600002604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600020201.mm.1 |
NM_001034902.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600020201.mm.1 |
NM_001034902.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600020202.mm.1 |
TC0600002604.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600020202.mm.1 |
NM_001034902.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600020202.mm.1 |
NM_001034902.2 |
Refseq |