| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25262927 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278335 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964339 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860243 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
488146 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
160911 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229831 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
927530 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
724966 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
682952 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
790976 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93231 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
722840 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851990 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401413 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
850594 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
531016 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396472 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404129 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
31797 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
638409 |
AK033679.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
459599 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
41362 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278390 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553327 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355352 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
AK005889.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
AK037364.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
AK047686.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
AK077339.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
AK084448.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
BC055955.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
334079 |
NM_181048.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
47514 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
103386 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
245913 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75916 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
41033 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174146 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97785 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
167081 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P157695 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262940 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262950 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262945 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262932 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262931 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262949 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262935 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262928 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262941 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262937 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262929 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262944 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262934 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262943 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25262946 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P157695 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06254614 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01557557 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01557576 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06254650 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P481316 |
BC150920.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P481316 |
NM_001037927.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P481316 |
AB205021.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06254650 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01557557 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P157695 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06162804 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06162810 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06162801 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06162798 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06162803 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25262927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263048 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263154 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263061 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262926 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263076 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263114 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263215 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262948 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263175 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263098 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263118 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263072 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263209 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263089 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263092 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263168 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263128 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262965 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263001 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263193 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262961 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263038 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263164 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263053 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262947 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262989 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262944 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262973 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263132 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263117 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25262929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263121 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25263146 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262931 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262945 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263164 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263161 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06493627 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262949 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263154 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262947 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262944 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262956 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06081730 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262952 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262968 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263158 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263156 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262934 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06183658 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262950 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262965 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262929 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06148366 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263169 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262935 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262941 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263170 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262946 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263152 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262937 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262928 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262958 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262961 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262954 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262940 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262948 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262967 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25262932 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06080395 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25263168 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06080394 |
- |
- |