Search
Search

Gm11437| Gene Symbol: Gm11437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037932.2 | NP_001033021.2 | 4 - 5 | 646 | 66 | |
| GenBank mRNA | AJ844653.1 | - | 4 - 5 | 646 | 66 | |
| BC147775.1 | - | 4 - 5 | 638 | 9056710 | 66 | |
| BC147791.1 | - | 4 - 5 | 638 | 9056714 | 66 | |
Set Membership: |
|