Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02745303_m1
          See other GM6994 GE Assays ›
          Gene Symbol:
          Gm6994
          Gene Name:
          predicted gene 6994
          Gene Aliases:
          EG629678
          Chromosome Location:
          Chr.14: 77479483 - 77506868 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02745303_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6994
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033141.1 4 - 5 376 80
          GenBank mRNA AY512927.1 - 4 - 5 376 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002457.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993640 NR_033141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993640 AY512927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993640 NR_033141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993640 AY512927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651101 NR_033141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651101 AY512927.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10421709 - -
          Affymetrix Mouse Clariom S Array PSR1400018953.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018953.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018953.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018950.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018950.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018950.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018951.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018951.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018951.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018952.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018952.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018952.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018954.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018954.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018954.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018957.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018957.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018957.mm.1 TC1400002457.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018955.mm.1 NR_033141.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018955.mm.1 NR_033141.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018955.mm.1 TC1400002457.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162379 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162379 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline