Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02745332_m1
          See other DEFB33 GE Assays ›
          Gene Symbol:
          Defb33
          Gene Name:
          defensin beta 33
          Gene Aliases:
          BD-33, EG654453
          Chromosome Location:
          Chr.8: 20892667 - 20897723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02745332_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039119.2 NP_001034208.1 2 - 3 210 69
          GenBank mRNA BC145764.1 - 1 - 2 148 40131067 69
          BC145766.1 - 1 - 2 148 40131069 69
          BY092288.1 - 2 - 3 211 69
          DQ012041.1 - 1 - 2 59 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000209.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179990 BC145766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179990 BY092288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179990 DQ012041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179990 NM_001039119.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2179990 BC145764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179990 BC145766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179990 BY092288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179990 DQ012041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179990 NM_001039119.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2179990 BC145764.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25705456 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577387 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577385 - -
          Affymetrix Mouse Clariom S Array PSR0800001781.mm.1 NM_001039119.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001781.mm.1 NM_001039119.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001781.mm.1 TC0800000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001782.mm.1 NM_001039119.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001782.mm.1 NM_001039119.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001782.mm.1 TC0800000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001780.mm.1 NM_001039119.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001780.mm.1 NM_001039119.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001780.mm.1 TC0800000209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001778.mm.1 NM_001039119.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001778.mm.1 NM_001039119.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001778.mm.1 TC0800000209.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161109 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161109 - -
          mouse-6 v1.1 (Illumina) GI_84993758-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161109 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline