Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02745496_s1
          See other GM6583 GE Assays ›
          Gene Symbol:
          Gm6583
          Gene Name:
          predicted gene 6583
          Gene Aliases:
          EG625424
          Chromosome Location:
          Chr.5: 112353773 - 112356033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02745496_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6583
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039228.4 NP_001034317.2 1 1078 65
          GenBank mRNA AK030210.1 - 1 1079 65
          BC150956.1 - 1 935 9087867 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P442118 NM_001039228.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P442118 AK030210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P442118 NM_001039228.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P442118 AK030210.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154844 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P442118 NM_001039228.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P442118 AK030210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P442118 NM_001039228.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P442118 AK030210.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23991666 - -
          Whole Mouse Genome G4122A A_51_P442118 NM_001039228.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442118 AK030210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442118 NM_001039228.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442118 AK030210.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23991720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04927550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04318411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04640744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04927548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04842043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23991657 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532535 - -
          Affymetrix Mouse Gene 1.1 ST Array 10532534 - -
          Affymetrix Mouse Clariom S Array PSR0500026765.mm.1 TC0500003015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026765.mm.1 NM_001039228.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026766.mm.1 TC0500003015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026766.mm.1 NM_001039228.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026764.mm.1 TC0500003015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026764.mm.1 NM_001039228.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026767.mm.1 TC0500003015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026767.mm.1 NM_001039228.4 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103060398 - -
          Illumina mouse-6 v1.0 expression beadchip 103060398 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2552338 - -
          Illumina Sentrix mouse-6.v1 bead array 103060398 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103060398 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2910876 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2552338 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2910876 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline