| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500025525.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025525.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025525.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025528.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025528.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025528.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025532.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025532.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025532.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025527.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025527.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025527.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025531.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025531.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025531.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025524.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025524.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025524.mm.1 |
NM_001039231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025526.mm.1 |
TC0500002892.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025526.mm.1 |
NM_001039231.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025526.mm.1 |
NM_001039231.3 |
Refseq |