| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10562655 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10562653 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552367 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10562654 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10562656 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552359 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10552362 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10562652 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10552358 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10562651 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10552363 |
- |
- |
| Mouse Genome 430 2.0 Array |
1443052_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1443052_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700026012.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026012.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026012.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026013.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026013.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026013.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026015.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026015.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026015.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026018.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026018.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026018.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026011.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026011.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026011.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026019.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026019.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026019.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026016.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026016.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026016.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026021.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026021.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026021.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026014.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026014.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026014.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026017.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026017.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026017.mm.1 |
NM_001242388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026022.mm.1 |
TC0700002931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700026022.mm.1 |
NM_001242388.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700026022.mm.1 |
NM_001242388.1 |
Refseq |
| Mouse Expression Set 430 |
1443052_PM_at |
- |
- |
| Mouse Expression Set 430 |
1443052_at |
- |
- |