Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02763057_u1
          See other CCL26 GE Assays ›
          Gene Symbol:
          Ccl26
          Gene Name:
          chemokine (C-C motif) ligand 26
          Gene Aliases:
          Ccl26l
          Chromosome Location:
          Chr.5: 135560448 - 135563569 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02763057_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013412.2 NP_001013430.2 3 239 138
          GenBank mRNA AY587128.1 - 3 138

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl26

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2175877 NM_001013412.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157041 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139675 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2175877 NM_001013412.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24116891 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139675 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24116858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04948136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04242970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04573683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04695033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04295231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04380713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04695035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04854944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24116892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534492 - -
          Affymetrix Mouse Gene 1.1 ST Array 10534489 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161570 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161570 - -
          mouse-6 v1.1 (Illumina) GI_61696139-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2905256 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2905256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jdch9:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0