Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02763059_g1
          See other TEX24 GE Assays ›
          Gene Symbol:
          Tex24
          Gene Name:
          testis expressed gene 24
          Gene Aliases:
          1700108N07Rik, TESF-1
          Chromosome Location:
          Chr.8: 27344323 - 27349188 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02763059_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013609.2 NP_001013627.1 2 1075 74
          GenBank mRNA AY552600.1 - 2 1075 74
          BC139071.1 - 2 1044 8862093 74
          BC139072.1 - 2 1044 8862094 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex24

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988364 NM_001013609.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988364 AK007143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988364 AY552600.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2495 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988364 NM_001013609.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988364 AK007143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988364 AY552600.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06458721 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06458727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25737757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10571169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571168 - -
          Affymetrix HT MG-430 PM Array Plate 1441893_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571167 - -
          Mouse Genome 430 2.0 Array 1441893_PM_at - -
          Mouse Genome 430 2.0 Array 1441893_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 541463_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 541463_at - -
          Affymetrix Mouse Clariom S Array PSR0800002722.mm.1 NM_001013609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002722.mm.1 NM_001013609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002722.mm.1 TC0800000357.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002725.mm.1 NM_001013609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002725.mm.1 NM_001013609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002725.mm.1 TC0800000357.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002723.mm.1 NM_001013609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002723.mm.1 NM_001013609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002723.mm.1 TC0800000357.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002726.mm.1 NM_001013609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002726.mm.1 NM_001013609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002726.mm.1 TC0800000357.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002721.mm.1 NM_001013609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002721.mm.1 NM_001013609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002721.mm.1 TC0800000357.mm.1 MTA TC
          Mouse Expression Set 430 1441893_PM_at - -
          Mouse Expression Set 430 1441893_at - -
          Murine Genome U74v2 Set 167924_at - -
          Murine Genome U74v2 Set 167924_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441893_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441893_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 808877 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2443237 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2785857 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2785857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline