| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1440342_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10607950 |
- |
- |
| Mouse Genome 430 2.0 Array |
1440342_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1440342_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0X00021102.mm.1 |
NR_029457.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021102.mm.1 |
NR_029457.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021102.mm.1 |
TC0X00003379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00021103.mm.1 |
NR_029457.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021103.mm.1 |
NR_029457.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021103.mm.1 |
TC0X00003379.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00021104.mm.1 |
NR_029457.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021104.mm.1 |
NR_029457.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00021104.mm.1 |
TC0X00003379.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1440342_at |
- |
- |
| Mouse Expression Set 430 |
1440342_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1440342_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1440342_PM_at |
- |
- |