Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02764733_s1
          See other RNF224 GE Assays ›
          Gene Symbol:
          Rnf224
          Gene Name:
          ring finger protein 224
          Gene Aliases:
          Gm757
          Chromosome Location:
          Chr.2: 25234476 - 25236787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02764733_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf224
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033410.2 NP_001028582.1 2 574 81
          GenBank mRNA AK146725.1 - 2 575 81
          BC147425.1 - 1 767 9056501 81
          BC147426.1 - 1 767 9056480 81

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045765 AK146725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045765 NM_001033410.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207111 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045765 AK146725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045765 NM_001033410.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070747 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04456932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01296042 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295993 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01813186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01813191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480648 - -
          Affymetrix Mouse Gene 1.1 ST Array 10480646 - -
          Affymetrix Mouse Clariom S Array PSR0200025802.mm.1 NM_001033410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025802.mm.1 NM_001033410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025802.mm.1 TC0200003138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025798.mm.1 NM_001033410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025798.mm.1 NM_001033410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025798.mm.1 TC0200003138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025799.mm.1 NM_001033410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025799.mm.1 NM_001033410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025799.mm.1 TC0200003138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025800.mm.1 TC0200003138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025801.mm.1 NM_001033410.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025801.mm.1 NM_001033410.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025801.mm.1 TC0200003138.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2782813 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2782813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline