Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02765631_s1
          See other ZFP868 GE Assays ›
          Gene Symbol:
          Zfp868
          Gene Name:
          zinc finger protein 868
          Gene Aliases:
          AI449175, AW108241, C530032E20, mszf62
          Chromosome Location:
          Chr.8: 69610654 - 69625562 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02765631_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp868
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045553.1 NP_001039018.1 5 1067 109
          NM_172754.2 NP_766342.2 5 1082 109
          GenBank mRNA AK083009.1 - 5 1049 109
          AK139223.1 - 5 1068 109
          AK168306.1 - 5 1072 109
          BC010277.1 - 4 686 3586128 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp868

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947613 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947617 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974855 NM_001045553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974855 NM_172754.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK168306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 BC010277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK083009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AB010369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK139223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK168306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 BC010277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK083009.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AB010369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P417990 AK139223.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45826 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102817 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106735 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK168306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 BC010277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK083009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AB010369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK139223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK168306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 BC010277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK083009.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AB010369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P417990 AK139223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974855 NM_001045553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974855 NM_172754.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25947565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06546430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06546443 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102817 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106735 - -
          Whole Mouse Genome G4122A A_52_P417990 AK168306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 BC010277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AK083009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AB010369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AK139223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AK168306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 BC010277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AK083009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AB010369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417990 AK139223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227463 AK168306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227463 BC010277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227463 AK083009.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227463 AK139223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P227463 NM_001045553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P351791 AK079745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P916422 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115951 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102817 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07195141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07195143 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07195142 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947613 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947617 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947615 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25947576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25947575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07197688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07108259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07195142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07197683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25947572 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579072 - -
          Affymetrix HT MG-430 PM Array Plate 1424091_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10579070 - -
          Mouse Genome 430 2.0 Array 1446529_PM_at - -
          Mouse Genome 430 2.0 Array 1424091_at - -
          Mouse Genome 430 2.0 Array 1424091_PM_at - -
          Mouse Genome 430 2.0 Array 1446529_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234362_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234362_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800017807.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017807.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017807.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017807.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017807.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017802.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017802.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017802.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017802.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017802.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017805.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017805.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017805.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017805.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017805.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017806.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017806.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017806.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017806.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017806.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017813.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017813.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017813.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017812.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017812.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017812.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017812.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017812.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017810.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017810.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017810.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017810.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017810.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017811.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017811.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017811.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017811.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017811.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017804.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017804.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017804.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017804.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017804.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017803.mm.1 NM_001045553.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017803.mm.1 NM_001045553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017803.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017803.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017803.mm.1 TC0800002368.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017814.mm.1 NM_172754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017814.mm.1 NM_172754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017814.mm.1 TC0800002368.mm.1 MTA TC
          Mouse Expression Set 430 1424091_PM_at - -
          Mouse Expression Set 430 1424091_at - -
          Murine Genome U74v2 Set 168304_r_at - -
          Murine Genome U74v2 Set 168304_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1424091_at - -
          Mouse Genome 430A 2.0 Array 1424091_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424091_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424091_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 487292 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440433 - -
          Illumina mouse-6 v1.0 expression beadchip 3440433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228048 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228048 - -
          mouse-6 v1.1 (Illumina) scl34728.6_247-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34728.6_247-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3143088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228048 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3143088 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline