Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02765657_sH
          See other CYPT2 GE Assays ›
          Gene Symbol:
          Cypt2
          Gene Name:
          cysteine-rich perinuclear theca 2
          Gene Aliases:
          Ckt1, Cypt2-ps
          Chromosome Location:
          Chr.X: 105499772 - 105500639 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02765657_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cypt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173436.2 NP_775612.2 2 555 95
          GenBank mRNA AF463499.1 - 2 538 95
          BC060953.1 - 2 555 6774045 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957539 NM_173436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957539 AF463499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957539 BC060953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 NM_173436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 AF463499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 BC060953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 NM_173436.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 AF463499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P135263 BC060953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 NM_173436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 AF463499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 BC060953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 NM_173436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 AF463499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P135263 BC060953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957539 NM_173436.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957539 AF463499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957539 BC060953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 NM_173436.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 AF463499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 BC060953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 NM_173436.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 AF463499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P135263 BC060953.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32580537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32580574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32580562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32580563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32580536 - -
          Whole Mouse Genome G4122A A_52_P135263 NM_173436.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P135263 AF463499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P135263 BC060953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P135263 NM_173436.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P135263 AF463499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P135263 BC060953.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32580563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14742385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15029723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32580578 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601348 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601349 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601347 - -
          Affymetrix Mouse Clariom S Array PSR0X00006992.mm.1 NM_173436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006992.mm.1 NM_173436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006992.mm.1 TC0X00001059.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00006993.mm.1 NM_173436.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006993.mm.1 NM_173436.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00006993.mm.1 TC0X00001059.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238134 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline