Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02765960_s1
          See other ZFP607 GE Assays ›
          Gene Symbol:
          Zfp607
          Gene Name:
          zinc finger proten 607
          Gene Aliases:
          4732475C15Rik
          Chromosome Location:
          Chr.7: 27859827 - 27909855 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02765960_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp607
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024726.2 NP_001019897.2 5 562 146
          GenBank mRNA AK028967.1 - 5 541 146
          AK143371.1 - 5 529 146
          AK163602.1 - 5 562 146
          BC042662.1 - 2218 146
          BC141161.1 - 5 529 9055773 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043662 AK163602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043662 NM_001024726.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043662 AK028967.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269405 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812047 AK030654.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93418 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107754 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169126 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043662 AK163602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043662 NM_001024726.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043662 AK028967.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25019188 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169126 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06148185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107754 - -
          Whole Mouse Genome G4122A A_52_P607586 BC141161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607586 AK143371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607586 AK163602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607586 NM_001024726.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P607586 AK028967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441999 BC141161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441999 AK163602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441999 NM_001024726.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P441999 AK028967.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120411 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169126 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25019202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06540690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06469358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06136546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06133574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06133573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06416038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25019271 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551433 - -
          Affymetrix Mouse Gene 1.1 ST Array 10551431 - -
          Affymetrix Mouse Clariom S Array PSR0700003856.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003856.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003856.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003853.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003853.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003853.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003860.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003860.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003860.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003859.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003859.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003859.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003854.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003854.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003854.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003857.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003857.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003857.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003858.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003858.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003858.mm.1 NM_001024726.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003861.mm.1 TC0700000476.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003861.mm.1 NM_001024726.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003861.mm.1 NM_001024726.2 Refseq
          Murine Genome U74v2 Set 129439_PM_at - -
          Murine Genome U74v2 Set 129439_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 487834 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106100673 - -
          Illumina mouse-6 v1.0 expression beadchip 106100673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217583 - -
          Illumina Sentrix mouse-6.v1 bead array 106100673 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106100673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2914036 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217583 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2914036 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0