Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03011142_m1
          See other PDXP GE Assays ›
          Gene Symbol:
          Pdxp
          Gene Name:
          pyridoxal (pyridoxine, vitamin B6) phosphatase
          Gene Aliases:
          1600027H05Rik, AB041662, PLPP
          Chromosome Location:
          Chr.15: 78913919 - 78919517 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03011142_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdxp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020271.3 NP_064667.2 1 - 2 613 122
          GenBank mRNA AB041662.1 - 1 - 2 508 122
          AK043228.1 - 1 - 2 574 122
          AY366300.1 - 1 - 2 575 122
          BC046955.1 - 1 - 2 564 122
          BC049107.1 - 1 - 2 613 122
          BC058388.1 - 1 - 2 159 122
          BC069982.1 - 1 - 2 385 122
          BU148105.1 - 1 - 2 382 122

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB041662.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdxp

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          regulation of mitotic nuclear division
          positive regulation of actin filament depolymerization
          actin rod assembly
          regulation of cytokinesis
          cellular response to ATP
          phosphoprotein phosphatase activity
          heat shock protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363154 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363148 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363152 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363150 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363147 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC069982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC046955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC049107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC058388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 AK043228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC069982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC046955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC049107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 BC058388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609120 AK043228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063686 BC069982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063686 BC046955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063686 BC049107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063686 BC058388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063686 AK043228.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387799 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC069982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC046955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC049107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC058388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 AK043228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC069982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC046955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC049107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 BC058388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609120 AK043228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063686 BC069982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063686 BC046955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063686 BC049107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063686 BC058388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2063686 AK043228.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363168 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08428849 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08428859 - -
          Whole Mouse Genome G4122A A_52_P609120 BC069982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC046955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC049107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC058388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 AK043228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC069982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC046955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC049107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 BC058388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609120 AK043228.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140826 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08428859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363150 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363152 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363147 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363154 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30363140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425160 - -
          Affymetrix HT MG-430 PM Array Plate 1417890_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581493 - -
          Affymetrix Mouse Gene 1.1 ST Array 10425158 - -
          Mouse Genome 430 2.0 Array 1417890_PM_at - -
          Mouse Genome 430 2.0 Array 1417890_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57028_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57028_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500004843.mm.1 NM_020271.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004843.mm.1 NM_020271.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004843.mm.1 TC1500002334.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500019339.mm.1 NM_020271.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019339.mm.1 NM_020271.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019339.mm.1 TC1500002334.mm.1 MTA TC
          Mouse Expression Set 430 1417890_PM_at - -
          Mouse Expression Set 430 1417890_at - -
          Mouse Genome 430A 2.0 Array 1417890_at - -
          Mouse Genome 430A 2.0 Array 1417890_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417890_at - -
          Affymetrix HT Mouse Genome 430A Array 1417890_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 785969 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228470 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228470 - -
          mouse-6 v1.1 (Illumina) scl47761.2_511-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47761.2_511-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228470 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228470 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl47761.2_511-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline