Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03011269_s1
          See other TAS2R140 GE Assays ›
          Gene Symbol:
          Tas2r140
          Gene Name:
          taste receptor, type 2, member 140
          Gene Aliases:
          T2R40, T2r34, TRB3, TRB5, Tas2r13, Tas2r137, Tas2r40, Tas2r8, mTRB3, mt2r64
          Chromosome Location:
          Chr.6: 133054855 - 133055793 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03011269_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r140
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021562.1 NP_067537.1 1 891 104
          GenBank mRNA BC145965.1 - 1 914 40131268 104
          BC145967.1 - 1 914 40131270 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r140

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 BC145965.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 BC145967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 NM_021562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 BC145965.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 BC145967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 NM_021562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360287 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 BC145965.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 BC145967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 NM_021562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 AF247735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 BC145965.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 BC145967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 NM_021562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 BC145965.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 BC145967.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 NM_021562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 BC145965.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 BC145967.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 NM_021562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24855884 - -
          Whole Mouse Genome G4122A A_51_P250571 BC145965.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 BC145967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 NM_021562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 AF247735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 BC145965.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 BC145967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 NM_021562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250571 AF247735.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24855883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05385012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05428296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05719684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24855884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05659037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548678 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548677 - -
          Affymetrix Mouse Clariom S Array PSR0600025841.mm.1 TC0600003292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025841.mm.1 NM_021562.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025841.mm.1 NM_021562.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025840.mm.1 TC0600003292.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100430546 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235837 - -
          mouse-6 v1.1 (Illumina) scl709.1.1_165-S - -
          Illumina Sentrix mouse-6.v1 bead array 100430546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100430546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2783117 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2783117 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235837 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline