Search
Search

2810474O19Rik| Gene Symbol: 2810474O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289661.1 | NP_001276590.1 | 5 - 6 | 5084 | 73 | |
| NM_001289662.1 | NP_001276591.1 | 4 - 5 | 4997 | 73 | ||
| NM_026054.3 | NP_080330.1 | 4 - 5 | 5015 | 73 | ||
| GenBank mRNA | AK220403.1 | - | 4 - 5 | 4998 | 73 | |
| BC055761.1 | - | 2 - 3 | 1086 | 73 | ||
| BC086679.1 | - | 2 - 3 | 4435 | 73 | ||
| BC089381.1 | - | 2 - 3 | 1033 | 73 | ||
| BC139435.1 | - | 4 - 5 | 4937 | 8862457 | 73 | |
| BC139436.1 | - | 5 - 6 | 5024 | 8862458 | 73 | |
Set Membership: |
|