Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03011963_g1
          See other UBE2S GE Assays ›
          Gene Symbol:
          Ube2s
          Gene Name:
          ubiquitin-conjugating enzyme E2S
          Gene Aliases:
          0910001J09Rik, 6720465F12Rik, AA409170, E2-EPF
          Chromosome Location:
          Chr.7: 4808014 - 4812340 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03011963_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2s
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133777.2 NP_598538.1 3 - 4 517 78
          GenBank mRNA AK003078.1 - 3 - 4 531 78
          AK017648.1 - 3 - 4 533 78
          AK020143.1 - 529 78
          AK146276.1 - 3 - 4 529 78
          AK154026.1 - 3 - 4 530 78
          AK154067.1 - 3 - 4 533 78
          BC012255.1 - 3 - 4 517 3581845 78
          BC030171.1 - 3 - 4 5037250 78
          BC083323.1 - 3 - 4 499 5715696 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ube2s

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ubiquitin-dependent protein catabolic process
          cell cycle
          exit from mitosis
          free ubiquitin chain polymerization
          anaphase-promoting complex-dependent catabolic process
          protein K29-linked ubiquitination
          protein K27-linked ubiquitination
          cell division
          protein K63-linked ubiquitination
          protein K11-linked ubiquitination
          protein K6-linked ubiquitination
          positive regulation of ubiquitin protein ligase activity
          nucleotide binding
          ubiquitin-protein transferase activity
          ATP binding
          ligase activity
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950631 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950636 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950639 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957168 BC030171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957168 BC083323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957168 AK146276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957168 AK154067.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957168 BC012255.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127459 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957168 BC030171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957168 BC083323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957168 AK146276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957168 AK154067.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957168 BC012255.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24950639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01475971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06114563 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06114586 - -
          Whole Mouse Genome G4122A A_51_P188782 BC030171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188782 BC083323.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188782 AK017648.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188782 AK146276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188782 AK154026.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140945 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137112 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140377 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01475971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06569558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06569565 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06569560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950637 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950631 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24950636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24950672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06573310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06423820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06569560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06491850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06229553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06544928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06377727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06487503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06515566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06229549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06569558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24950625 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559678 - -
          Affymetrix HT MG-430 PM Array Plate 1430962_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416726_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559676 - -
          Affymetrix Mouse Gene 1.1 ST Array 10585586 - -
          Mouse Genome 430 2.0 Array 1416726_PM_s_at - -
          Mouse Genome 430 2.0 Array 1430962_at - -
          Mouse Genome 430 2.0 Array 1430962_PM_at - -
          Mouse Genome 430 2.0 Array 1416726_s_at - -
          Affymetrix Mouse Clariom S Array PSR0700019857.mm.1 TC0700002173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019857.mm.1 NM_133777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019857.mm.1 NM_133777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019858.mm.1 TC0700002173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019858.mm.1 NM_133777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019858.mm.1 NM_133777.2 Refseq
          Mouse Expression Set 430 1430962_PM_at - -
          Mouse Expression Set 430 1416726_s_at - -
          Mouse Expression Set 430 1430962_at - -
          Mouse Expression Set 430 1416726_PM_s_at - -
          Murine Genome U74v2 Set 94274_PM_at - -
          Murine Genome U74v2 Set 94274_at - -
          Murine Genome U74v2 Set 171178_PM_r_at - -
          Murine Genome U74v2 Set 171178_r_at - -
          Mouse Genome 430A 2.0 Array 1430962_PM_at - -
          Mouse Genome 430A 2.0 Array 1430962_at - -
          Mouse Genome 430A 2.0 Array 1416726_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1416726_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416726_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430962_at - -
          Affymetrix HT Mouse Genome 430A Array 1416726_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430962_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 498922 - -
          ABI Mouse Genome Survey Microarray 843907 - -
          ABI Mouse Genome Survey Microarray 921013 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2424400 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2424400 - -
          mouse-6 v1.1 (Illumina) scl31799.8.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31799.8.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2424399 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2424399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline