Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03012618_s1
          See other OLFR147 GE Assays ›
          Gene Symbol:
          Olfr147
          Gene Name:
          olfactory receptor 147
          Gene Aliases:
          M3, MOR164-1
          Chromosome Location:
          Chr.9: 38401711 - 38403829 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03012618_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr147
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146869.2 NP_667080.1 2 1119 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr147

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2146037 NM_146869.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173981 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146037 NM_146869.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434612 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173981 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118178 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173981 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07688554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07688556 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07688548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07491783 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07491767 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07491774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07511184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434570 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584406 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584405 - -
          Affymetrix Mouse Clariom S Array PSR0900002797.mm.1 NM_146869.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002797.mm.1 NM_146869.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002797.mm.1 TC0900000398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900002795.mm.1 NM_146869.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002795.mm.1 NM_146869.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002795.mm.1 TC0900000398.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2533404 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793865 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline