| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1960251 |
BC054854.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
567275 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
56329 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
124427 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
304608 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758699 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
210246 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853114 |
AF215892.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853114 |
AK013647.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853114 |
BC048471.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853114 |
NM_021452.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853114 |
XM_001480418.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
736579 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924445 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347417 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
553928 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785349 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
422538 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
499790 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
218452 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
716560 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855273 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299593 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326942 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577368 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
537392 |
AK015450.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
33342 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
5765 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
703552 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396482 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
835533 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
347222 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
527765 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
60962 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
164870 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
178995 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
112671 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
154038 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176598 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153632 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
139502 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P113385 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130926 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1960251 |
BC054854.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726786 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726801 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726766 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726784 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726789 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726794 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726781 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726770 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726771 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726791 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726787 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726769 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726785 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726797 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726778 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726775 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30726798 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585473 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585454 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585539 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130926 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585536 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03185816 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585516 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P113385 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08585504 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P560176 |
BC054854.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P560176 |
NM_001005854.2 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P113385 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08585536 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03185815 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08585554 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130926 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12651490 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12700553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726791 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726695 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726693 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726574 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726587 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726590 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726537 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726621 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726647 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726689 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726674 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726789 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726767 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726570 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726684 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726588 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726810 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726534 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726682 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726680 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726667 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726576 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726527 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726571 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726626 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726531 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726606 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726525 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726530 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726523 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726663 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726683 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726729 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726567 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726584 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726703 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726641 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726645 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726784 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726529 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726677 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726578 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726550 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726554 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726536 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726711 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726552 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726758 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726539 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30726734 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726801 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726802 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726626 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726798 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726612 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726807 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726758 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726809 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726759 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726780 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726604 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726794 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12545653 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726621 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726784 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726775 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726773 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726760 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726767 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726782 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726787 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726771 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726786 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726610 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12545661 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12545666 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726756 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726768 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726789 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726774 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726605 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726606 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726625 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726762 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726627 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726601 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726796 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726805 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12919841 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726616 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12682472 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726785 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726597 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12545683 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726778 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726614 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30726791 |
- |
- |