Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03013476_s1
          See other OLFR225 GE Assays ›
          Gene Symbol:
          Olfr225
          Gene Name:
          olfactory receptor 225
          Gene Aliases:
          MOR256-51
          Chromosome Location:
          Chr.11: 59610786 - 59614225 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03013476_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr225
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011740.2 NP_001011740.2 1 945 83

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr225

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266876 NM_001011740.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266876 NM_001011740.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266876 NM_001011740.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266876 NM_001011740.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266876 NM_001011740.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266876 NM_001011740.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27837888 - -
          Whole Mouse Genome G4122A A_51_P266876 NM_001011740.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266876 NM_001011740.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405737 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09516300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09216866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09060502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09516291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09196382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09216862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09706486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09027717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837867 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376533 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376532 - -
          Affymetrix Mouse Clariom S Array PSR1100005808.mm.1 NM_001011740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005808.mm.1 NM_001011740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005808.mm.1 TC1100000727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005809.mm.1 NM_001011740.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005809.mm.1 NM_001011740.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005809.mm.1 TC1100000727.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160900 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160900 - -
          mouse-6 v1.1 (Illumina) GI_58801273-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160900 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160900 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0