Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03013516_s1
          See other OLFR1089 GE Assays ›
          Gene Symbol:
          Olfr1089
          Gene Name:
          olfactory receptor 1089
          Gene Aliases:
          MOR193-1
          Chromosome Location:
          Chr.2: 86732675 - 86733610 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03013516_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1089
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011771.2 NP_001011771.2 1 901 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003955.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1089

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977446 NM_001011771.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977446 NM_001011771.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00425004 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00425004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21403434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01677894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01677896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01500736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21403438 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484629 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484628 - -
          Affymetrix Mouse Clariom S Array PSR0200033806.mm.1 NM_001011771.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033806.mm.1 NM_001011771.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033806.mm.1 TC0200003955.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161187 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161187 - -
          mouse-6 v1.1 (Illumina) GI_58801319-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925855 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925855 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0