Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03014045_m1
          See other ZFP867 GE Assays ›
          Gene Symbol:
          Zfp867
          Gene Name:
          zinc finger protein 867
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 59461197 - 59472566 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03014045_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp867
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178417.3 NP_848504.2 3 - 4 323 103
          GenBank mRNA AK132062.1 - 3 - 4 324 103
          BC050078.1 - 3 - 4 302 30008012 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002852.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27837109 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27837111 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123047 AK132062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123047 NM_178417.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2363442 AK132062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2363442 BC050078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2363442 NM_178417.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133774 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154798 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176210 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2363442 AK132062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2363442 BC050078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2363442 NM_178417.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123047 AK132062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123047 NM_178417.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154798 - -
          Whole Mouse Genome G4122A A_52_P334808 AK132062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334808 BC050078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334808 NM_178417.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176210 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09387074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09387079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27837109 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27837111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27836990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27836999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27836992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27836997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27836991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27837053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09284848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09387071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09387078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09284852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09344659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09198589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09014640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09519205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09485841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09245218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09284849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27837111 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10386396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10386395 - -
          Affymetrix HT MG-430 PM Array Plate 1438079_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10386394 - -
          Mouse Genome 430 2.0 Array 1438079_at - -
          Mouse Genome 430 2.0 Array 1438079_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237775_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237775_at - -
          Affymetrix Mouse Clariom S Array PSR1100025873.mm.1 NM_178417.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025873.mm.1 NM_178417.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025873.mm.1 TC1100002852.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025874.mm.1 NM_178417.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025874.mm.1 NM_178417.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025874.mm.1 TC1100002852.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025876.mm.1 NM_178417.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025876.mm.1 NM_178417.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025876.mm.1 TC1100002852.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025877.mm.1 NM_178417.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025877.mm.1 NM_178417.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025877.mm.1 TC1100002852.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100025875.mm.1 NM_178417.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025875.mm.1 NM_178417.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100025875.mm.1 TC1100002852.mm.1 MTA TC
          Mouse Expression Set 430 1438079_at - -
          Mouse Expression Set 430 1438079_PM_at - -
          Murine Genome U74v2 Set 167922_PM_r_at - -
          Murine Genome U74v2 Set 167922_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1438079_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438079_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232229 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232229 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline