Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03014429_m1
          See other SERPINB3D GE Assays ›
          Gene Symbol:
          Serpinb3d
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 3D
          Gene Aliases:
          Serpinb3l1
          Chromosome Location:
          Chr.1: 107078163 - 107084125 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03014429_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb3d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201376.1 NP_958764.1 2 - 3 219 147
          GenBank mRNA AY367776.1 - 2 - 3 219 147
          BC138290.1 - 2 - 3 245 8861309 147
          BC138291.1 - 2 - 3 245 8861310 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002904.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinb3d

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          negative regulation of peptidase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of endopeptidase activity
          negative regulation of endopeptidase activity
          positive regulation of cell migration
          autocrine signaling
          paracrine signaling
          negative regulation of catalytic activity
          negative regulation of JUN kinase activity
          negative regulation of proteolysis
          viral entry into host cell
          virus receptor activity
          protease binding
          serine-type endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159791 BC138290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159791 BC138291.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159791 NM_201376.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159791 AY367776.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219565 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159791 BC138290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159791 BC138291.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159791 NM_201376.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159791 AY367776.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20511678 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04217453 - -
          Whole Mouse Genome G4122A A_52_P565455 BC138290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P565455 BC138291.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P565455 NM_201376.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P565455 AY367776.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00160124 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00160121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20511628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01021793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00160126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00160124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00178462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20511692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357079 - -
          Affymetrix Mouse Gene 1.1 ST Array 10357077 - -
          Affymetrix Mouse Clariom S Array PSR0100023491.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023491.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023491.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023497.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023497.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023497.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023498.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023496.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023496.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023496.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023490.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023495.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023495.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023495.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023492.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023492.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023492.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023493.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023493.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023493.mm.1 TC0100002904.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100023494.mm.1 NM_201376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023494.mm.1 NM_201376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023494.mm.1 TC0100002904.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 787599 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5670072 - -
          Illumina mouse-6 v1.0 expression beadchip 5670072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256750 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256750 - -
          mouse-6 v1.1 (Illumina) scl0394252.4_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0394252.4_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009747 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256750 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009747 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline